Learn More
Medchemexpress LLC Tcmdc-135051 hydrochloride | 2705545-47-5 | 98.2% | 508.05 | C29H34ClN3O3 | 10MG

Supplier: Medchemexpress LLC HY126323B10MG
TCMDC-135051 hydrochloride is the hydrochloride salt of a selective PfCLK3 inhibitor used in antimalarial research. It is supplied as a light yellow to yellow solid with a molecular weight of 508.05 g/mol and a reported purity of 98.17%.
- Light yellow to yellow solid physical form.
- Reported purity 98.17%.
- Molecular formula C29H34ClN3O3 and molecular weight 508.05 g/mol.
- Available in small research pack sizes, including 10 mg.
- Store solid at 4°C, sealed, away from moisture; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.