Learn More
Medchemexpress LLC TCMDC-135051 hydrochloride | 2705545-47-5 | 98.2% | 508.05 | C29H34ClN3O3 | 50MG
SDP
Supplier:  Medchemexpress LLC HY126323B50MG
Encompass_Preferred
TCMDC-135051 hydrochloride is the hydrochloride salt of a selective PfCLK3 inhibitor supplied for research use. It is provided as a dry solid and as a DMSO solution for in vitro assays, with reported DMSO solubility of 83.33 mg/mL.
Catalog No. 50-002-36734