Learn More
Selleck Chemical LLC TCN 201 S0446-25MG
SDP
Supplier:  Selleck Chemical LLC S044625MG
Encompass_Preferred
TCN-201 is a selective GluN1/GluN2A (NR1/NR2A) NMDA receptor antagonist with pIC50 6 8 for NR1/NR2A and 4 3 for NR1/NR2B (GluN1/GluN2B) respectively
Catalog No. 50-000-42327