Learn More
Medchemexpress LLC Tcn 213 | 556803-08-8 | MFCD04788042 | 99.2% | 376.54 g/mol | C18H24N4OS2 | 100 MG

Supplier: Medchemexpress LLC HY107712100MG
TCN 213 is a selective, glycine-dependent GluN1/GluN2A NMDA receptor antagonist used as a research chemical for neuropharmacology and receptor characterization. It is supplied as a solid with high reported purity and is typically prepared in DMSO for in vitro assays and mechanistic studies.
- Selective GluN1/GluN2A NMDA receptor antagonist suitable for receptor pharmacology studies.
- Reported purity 99.2% supports reproducible experimental results.
- Available in small-molecule research sizes for assay development and testing.
- Soluble in DMSO for preparation of concentrated stock solutions.
- Molecular weight 376.54 g/mol and formula C18H24N4OS2 for stoichiometric calculations.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.