Learn More
Medchemexpress LLC TCO-NHS Ester (axial) | 1610931-22-0 | C13H17NO5 | 5 MG

Supplier: Medchemexpress LLC HY1516985MG
TCO-NHS Ester axial is a click chemistry reagent that can be used as a building block in amine reactions. It contains TCO groups, which can undergo inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing Tetrazine groups.
- Click chemistry reagent.
- Can be used as a building block in amine reactions.
- Contains TCO groups for inverse electron demand Diels-Alder reaction (iEDDA) with tetrazine groups.
- Solid appearance.
- White to off-white color.
- Purity of 98.0% or greater.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.