Learn More
Medchemexpress LLC TCO-PEG3-Biotin | 570.74 | C27H46N4O7S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1360505MG
Encompass_Preferred
TCO-PEG3-Biotin is a cleavable three-unit polyethylene glycol (PEG) linker bearing a trans-cyclooctene (TCO) group and a biotin tag used for click chemistry and antibody-drug conjugate (ADC) synthesis. It enables rapid inverse electron-demand Diels-Alder (iEDDA) ligation with tetrazine partners and supports affinity capture or detection via the biotin handle.
  • Cleavable 3-unit PEG linker for bioconjugation.
  • Trans-cyclooctene (TCO) group enables inverse electron-demand Diels-Alder (iEDDA) click reactions.
  • Biotin tag facilitates affinity capture and detection workflows.
  • Molecular weight 570.74 and formula C27H46N4O7S support stoichiometric conjugation planning.
  • Available in small milligram quantities suitable for conjugation development and testing.
Catalog No. 50-002-02697