Learn More
Medchemexpress LLC TCO-PEG3-Biotin | 99.9% | 570.74 g/mol | C27H46N4O7S | 10 MG

Supplier: Medchemexpress LLC HY13605010MG
TCO-PEG3-Biotin is a cleavable three-unit PEG linker used for antibody-drug conjugate (ADC) synthesis and click-chemistry bioconjugation. It contains a trans-cyclooctene (TCO) moiety that reacts with tetrazine-functionalized partners via inverse electron-demand Diels-Alder (iEDDA) chemistry. This product is for research use only.
- Cleavable three-unit PEG linker suitable for ADC synthesis.
- Enables rapid iEDDA click chemistry with tetrazine-functionalized molecules.
- High purity: 99.9%.
- Molecular weight: 570.74 g/mol.
- Chemical formula: C27H46N4O7S.
- Appearance: white to off-white solid.
- Storage: powder at -20°C; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
- Available quantity: 10 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.