Learn More
Medchemexpress LLC TCO-PEG3-maleimide | 1809356-72-6 | C26H41N3O8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1516965MG
Encompass_Preferred
TCO-PEG3-maleimide is a click chemistry reagent that efficiently binds the TCO (trans-cyclooctene) moiety to thiol-containing molecules. This reagent is ideal for research applications involving antibodies and cysteine-containing peptides.
  • Click chemistry reagent
  • Binds TCO moiety to thiol-containing molecules
  • Suitable for antibodies
  • Suitable for cysteine-containing peptides
  • For research use only
Catalog No. 50-003-72939