Learn More
Medchemexpress LLC TCO-PEG4-NHS ester | 1621096-79-4 | 99.2% | C24H38N2O10 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141167A100MG
Encompass_Preferred
TCO-PEG4-NHS ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It also functions as a click chemistry reagent, featuring a TCO group that facilitates an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing Tetrazine groups. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Features a TCO group for iEDDA reactions
  • Reacts with molecules containing Tetrazine groups
Catalog No. 50-003-54900