Learn More
Medchemexpress LLC TCO-PEG4-NHS ester | 1621096-79-4 | MFCD28118912 | 95.4% | 514.57 | C24H38N2O10 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY141167A5MG
Encompass_Preferred
TCO-PEG4-NHS ester is a bifunctional PEG-based linker combining a trans-cyclooctene (TCO) group and an N-hydroxysuccinimide (NHS) ester for efficient bioorthogonal conjugation. It is provided as a high-purity reagent for click chemistry and PROTAC linker synthesis, compatible with protein and antibody labeling in common organic solvents and aqueous buffers.
  • Bifunctional linker with trans-cyclooctene and NHS ester for rapid bioorthogonal conjugation.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • High purity suitable for analytical and preparative work.
  • Compatible with protein, antibody, and small-molecule labeling workflows.
  • Supplied in small quantities for research-scale applications.
Catalog No. 50-002-22783