Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tco-peg4-vc-pab-mmae | 2758671-45-1 | 99.3% | 1522.91 | C78H127N11O19 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1480571MG

Encompass_Preferred

TCO-PEG4-VC-PAB-MMAE is a drug-linker conjugate used in antibody-drug conjugate (ADC) research and click-chemistry applications; it couples a trans-cyclooctene (TCO) reactive handle to a PEG4-spaced valine-citrulline-para-aminobenzyl (Val-Cit-PAB) cleavable linker and the cytotoxic payload MMAE.

  • High purity (99.3%).
  • Molecular weight 1522.91 g/mol.
  • Chemical formula C78H127N11O19.
  • CAS number 2758671-45-1.
  • Suitable for iEDDA click chemistry with tetrazine reagents.
  • Intended for ADC synthesis and conjugation research.
  • Store at -20°C and protect from light; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
  • Available in small research pack sizes (e.g., 1 MG).

Catalog No. 50-002-03405


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.