Learn More
Medchemexpress LLC Tco4-peg7-maleimide | 99.0% | 671.78 g/mol | C32H53N3O12 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15639210MG
Encompass_Preferred
TCO4-PEG7-Maleimide is a PEG7-based PROTAC linker that combines a trans-cyclooctene (TCO) moiety with a maleimide reactive handle. It is formulated for bioconjugation and the construction of PROTACs or other conjugates using orthogonal click chemistry, and is supplied in research-scale quantities as a colorless to light-yellow oil.
  • Enables strain-promoted click reactions with tetrazine-functionalized partners.
  • Reacts selectively with thiol groups via maleimide chemistry for cysteine conjugation.
  • PEG7 spacer improves solubility and provides controlled linker length.
  • High purity suitable for research applications.
  • Soluble in DMSO; ultrasonic assistance may be needed for dissolution.
Catalog No. 50-002-88080