Learn More
Beckman Coulter TCR Vbeta13.1-PE, IMMU 222, 1 mL, ASR
Supplier:  Beckman Coulter IM2292
Encompass
Also called TCRBV13S1 V beta 13 is a complex subfamily of the T cell receptor. V beta 13.1 is expressed by 1.8% to 3.3% of peripheral CD3+ cells in normal blood. This sequence is also referred to as TRBV6-5, TRBV6-6, TRBV6-9 (based on the IMGT gene nomenclature).
Catalog No. NC1909485