Learn More
Abcam TCS 359, FLT3 receptor tyrosine kinase inhibitor, 50MG
Supplier:  Abcam AB14126850MG
Encompass_Preferred
MW 360.4 Da, Purity >99%. Potent FLT3 receptor tyrosine kinase inhibitor (IC₅₀ = 42 nM). Selective over 22 other kinases. Shows antiproliferative effects (IC₅₀ = 340 nM for Human acute myelocytic leukemia MV4-11 cells).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6737