Learn More
Abcam TCS Antibody Purification Kit, 3 Tests
Supplier:  Abcam AB1092073TEST
Encompass_Preferred
TCS Antibody Purification Kit (ab109207) has been designed for tissue culture supernatant purification, and can be used to purify 10-50 mL in each purification.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5213