Learn More
Abcam TCS O x 2 29, Ore x in-2 receptor antagonist, 50MG
Supplier:  Abcam AB14131650MG
Encompass_Preferred
MW 434 Da, Purity >98%. Potent, selective orexin-2 receptor antagonist (IC₅₀ = 40 nM). Neurological and insomnia agent. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6684