Learn More
Apexbio Technology LLC TCS-PIM-1-4a 327033-36-3 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B47645.1
Encompass_Preferred
TCS-PIM-1-4a is a small-molecule inhibitor targeting Pim kinases a family of serine/threonine kinases It is designed to selectively inhibit Pim kinases thereby modulating signaling pathways related to cell growth and survival TCS-PIM-1-4a exerts its biological activity primarily through activation of AMP-activated protein kinase (AMPK) resulting in inhibition of the mammalian target of rapamycin complex 1 (mTORC1) signaling pathway In cell-based studies TCS-PIM-1-4a demonstrates cytotoxic activity against diverse hematological tumor cell lines including myeloid and lymphoid subtypes with IC50 values ranging from approximately 0 8 to 40 M Based on these pharmacological properties TCS-PIM-1-4a holds research potential in investigations of Pim kinase signaling and therapeutic strategies for hematological malignancies
Catalog No. 50-000-89513