Learn More
Medchemexpress LLC TCV-309 chloride | 121494-09-5 | 95.0% | 629.97 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY19121A10MM/1ML
Encompass_Preferred
TCV-309 chloride is a potent and specific platelet activating factor (PAF) antagonist. It specifically inhibits PAF-induced aggregation of rabbit and human platelets, and [3H]PAF binding to rabbit platelet microsomes with IC50 values of 33 nM, 58 nM and 27 nM, respectively. TCV-309 chloride has beneficial effects in anaphylactic shock. For research use only.
  • Potent and specific platelet activating factor (PAF) antagonist.
  • Inhibits PAF-induced aggregation of rabbit and human platelets.
  • Inhibits [3H]PAF binding to rabbit platelet microsomes.
  • Beneficial effects in anaphylactic shock.
  • Selectively inhibits PAF-induced hypotension, hemoconcentration, and death in rats.
  • Most potently protects mice from death induced by PAF and due to anaphylactic shock.
  • Reversed PAF-induced hypotension and endotoxin-induced hypotension in rats.
Catalog No. 50-003-85800