Learn More
Medchemexpress LLC TD-0212 TFA 25mg | 1073549-11-7 | 25 MG

Supplier: Medchemexpress LLC HY114412A25MG
TD-0212 TFA is the trifluoroacetic acid (TFA) salt of TD-0212, an orally active small molecule with dual activity as an angiotensin II type 1 (AT1) receptor antagonist and a neprilysin (NEP) inhibitor. It is supplied as a white to off-white solid for research use.
- CAS number: 1073549-11-7.
- Dual pharmacology: AT1 antagonist (pKi 8.9) and NEP inhibitor (pIC50 9.2).
- Molecular formula: C30H35F4N3O6S; molecular weight: 641.67 g/mol.
- Purity: 98.44%.
- Solubility: DMSO 125 mg/mL; in vivo vehicle protocols achieve ≥2.08 mg/mL.
- Storage: solid at -20°C; in solvent -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.