Learn More
Medchemexpress LLC HY-114406 10mg Medchemexpress, TD-106 CAS:2250288-69-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11440610MG
Encompass_Preferred
Medchemexpress, HY-114406 10mg TD-106 CAS:2250288-69-6 TD-106 is a cereblon (CRBN) modulator, which can be used for targeted protein degradation. BRD4 PROTACs with TD-106 induce BRD4 degradation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9347