Learn More
Medchemexpress LLC TD-165 | 2305936-56-3 | >98.1% | 50 MG

Supplier: Medchemexpress LLC HY13071450MG
TD-165 is a PROTAC-based cereblon (CRBN) degrader comprising a cereblon (CRBN) ligand binding group, a linker, and a von Hippel-Landau (VHL) binding group. This compound is intended for research use only.
- Demonstrates a 50% degradation concentration (DC50) of 20.4 nM.
- Achieves a maximum degradation (Dmax) value of 99.6%.
- Leads to a concentration-dependent decrease in CRBN proteins in HEK293T cells.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.