Learn More
TARGETMOL CHEMICALS INC TD139 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T51215MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. TD139 is an inhaled galectin-3 inhibitor (Kd 14 nM). purity: 99%
Catalog No. 50-257-9810