Learn More
Strem, An Ascensus Company CAS# [1066-77-9]. 25g. Tetrakis(dimethylamino)tin(IV), 99% (99.99%-Sn) TDMASn PURATREM, 50-1815, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00014860
SDP
Supplier:  Strem, An Ascensus Company 98405025G
Encompass
CAS# [1066-77-9]. 25g. Tetrakis(dimethylamino)tin(IV), 99% (99.99%-Sn) TDMASn PURATREM, 50-1815, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00014860. Molecular Weight: 295.01. Molecular Formula: Sn[N(CH3)2]4. Color/form: colorless to pale-yellow liq. Strem# 98-4050. www.strem.com/catalog/v3/98-4050/
Catalog No. 50-197-9400