Learn More
Selleck Chemical LLC TDZD-8 S2926-5mg
SDP
Supplier:  Selleck Chemical LLC S29265MG
Encompass_Preferred
TDZD-8 (NP 01139) is a non-ATP competitive GSK-3 inhibitor with IC50 of 2 M minimal inhibitory effect observed on CDK1 casein kinase II PKA and PKC
Catalog No. 50-311-2492