Learn More
AdipoGen TDZD-8 (5 mg)
SDP
Supplier:  AdipoGen AGCR13740M005
Encompass_Preferred
Chemical. CAS: 327036-89-5. Formula: C10H10N2O2S. MW: 222.3. TDZD-8 is a non-ATP competitive selective inhibitor of the serine/threonine kinase glycogen synthase kinase 3beta (GSK3beta; IC50 = 2µM), binding to the active site of GSK3beta. It does not significantly affect the activities of Cdk-1/cyclin B, CK-II, cAMP-dependent protein kinase or protein kinase C (IC50s = >100 µM), but inhibits PKC and Flt-3 activities (IC50 = 1.4-5.5µM and 673nM, respectively). TDZD-8 is a potent anti-proliferative and pro-apoptotic agent of glioma cells in vitro and in vivo. These effects are associated with an early activation of extracellular signal-regulated kinase (ERK), which is followed by an increased expression of the early growth response-1 (EGR-1) and p21. TDZD-8 potentially inhibits the 33.8-kDa Main protease (Mpro)/3C-like protease of SARS-CoV-2, consequently inhibiting viral transcription and replication and possibly inhibiting spread of COVID-19.
Catalog No. 50-205-4170