Learn More
Teknova TE Buffer with 0.1 mM EDTA, pH 7.6, RNAse/DNAse Tested, 500mL, Sterile. 4 pack.
SDP
Supplier:  Teknova T02304PK
Encompass
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.
Catalog No. 50-238-7349