Learn More
Teknova DNA Suspension Buffer, pH 8.0. 100mL, Sterile. DNase/RNase Tested, PCR Grade.
SDP
Supplier:  Teknova T0227
Encompass
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.
Catalog No. 50-843-207