Learn More
Medchemexpress LLC Tead-in-8 | 1008768-41-9 | 99.8% | 435.56 | C26H33N3O3 | 10 MG

Supplier: Medchemexpress LLC HY16317310MG
TEAD-IN-8 is a small-molecule TEAD inhibitor that potently inhibits TEAD-YAP transcriptional activity and shows antiproliferative effects in YAP-dependent cancer cell models.
- Novel TEAD inhibitor mechanism targeting TEAD-YAP transcriptional activity.
- Demonstrates antiproliferative effects in YAP-dependent cancer cells.
- High purity (99.8%) suitable for research applications.
- Molecular weight 435.56 g/mol; formula C26H33N3O3.
- Available in multiple pack sizes, including 10 MG.
- Storage: powder -20°C (stable for years) and 4°C (short-term); in solvent -80°C or -20°C per guidance.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.