Learn More
Medchemexpress LLC Tebanicline dihydrochloride | 209326-19-2 | 99.5% | C9H13Cl3N2O | 10MG
SDP
Supplier:  Medchemexpress LLC HY14316A10MG
Encompass_Preferred
Tebanicline dihydrochloride is the dihydrochloride salt of tebanicline (ebanicline/ABT-594), a neuronal nicotinic acetylcholine receptor agonist reported to have potent analgesic activity. Supplied as a research-grade solid with supplier characterization and safety documentation, it is intended for in vitro and in vivo research where well-defined pharmacological activity is required.
  • High purity suitable for research applications.
  • Solid, white to off-white appearance for easy handling.
  • Molecular formula C9H13Cl3N2O and molecular weight 271.57 g/mol.
  • Available in multiple small pack sizes for dose-ranging studies.
  • Provided with supplier datasheet and safety documentation.
Catalog No. 50-002-61338