Learn More
Medchemexpress LLC Tebanicline (dihydrochloride) | 209326-19-2 | MFCD01758464 | 99.5% | 271.57 g/mol | C9H13Cl3N2O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY14316A5MG
Encompass_Preferred
Tebanicline dihydrochloride is the dihydrochloride salt of tebanicline (ABT-594), a potent nicotinic acetylcholine receptor (nAChR) ligand used in research on analgesia and receptor pharmacology. The compound is supplied as a solid research-use reagent (CAS 209326-19-2) with molecular formula C9H13Cl3N2O and molecular weight 271.57 g/mol.
Catalog No. 50-002-21455