Learn More
Medchemexpress LLC Tebentafusp 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP993391MG
Encompass_Preferred
Tebentafusp (IMCgp100) is a bispecific fusion protein to target gp100 peptide-HLA-A 02 01 (a melanoma-associated antigen) Tebentafusp guides T cells to kill gp100-expressing tumor cells via a high affinity T-cell receptor (TCR) binding domain and an anti-CD3 T-cell engaging domain Tebentafusp leads to inflammatory cytokines and cytolytic proteins production resulting in the direct lysis of tumour cells[1][2]
Catalog No. 50-003-23941