Learn More
Medchemexpress LLC Tebipenem pivoxil | 161715-24-8 | 99.7% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0396100MG
Encompass_Preferred
Tebipenem pivoxil (L084) is an orally active antibiotic that targets a broad spectrum of pathogenic bacteria. It functions by binding to penicillin-binding protein (PBP), which inhibits bacterial cell wall synthesis. This mechanism makes it a promising agent for treating various infections.
  • Orally active antibiotic
  • Effective against a variety of pathogenic bacteria
  • Binds to penicillin-binding protein (PBP)
  • Inhibits bacterial cell wall synthesis
  • Demonstrates excellent antibacterial activity with MIC50s below 64 μg/mL against tested Gram-positive and Gram-negative bacteria in vitro
  • In vivo studies show significant protection for sepsis mice challenged with various pathogenic bacteria
Catalog No. 50-004-06512