Learn More
Medchemexpress LLC Tebipenem pivoxil | 161715-24-8 | 99.7% | 497.63 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHYB0396
Encompass_Preferred
Tebipenem pivoxil is an orally active antibiotic that works against various pathogenic bacteria. It functions by binding to penicillin-binding protein (PBP), thereby inhibiting bacterial cell wall synthesis. It exhibits excellent antibacterial activity against a range of pathogenic bacteria.
  • Orally active antibiotic
  • Inhibits cell wall synthesis by binding to penicillin-binding protein (PBP)
  • Exhibits excellent antibacterial activity against pathogenic bacteria
  • Inhibits Gram-positive and Gram-negative bacteria with MIC50s below 64 μg/mL
  • Significantly increased survival in sepsis mouse models
Catalog No. 50-003-65562