Learn More
Medchemexpress LLC Tebipenem pivoxil | 161715-24-8 | 99.7% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB039650MG
Encompass_Preferred
Tebipenem pivoxil (L084) is an orally active antibiotic that effectively targets various pathogenic bacteria by inhibiting cell wall synthesis through binding to penicillin-binding protein (PBP). It is intended for research use only.
  • Orally active antibiotic
  • Effective against various pathogenic bacteria
  • Inhibits cell wall synthesis
  • Binds to penicillin-binding protein (PBP)
  • Shows excellent antibacterial activity (0-128 μg/mL; 18-24 h)
  • Inhibits Gram-positive and Gram-negative bacteria with MIC50s below 64 μg/mL
Catalog No. 50-004-06833