Learn More
Medchemexpress LLC Tebipenem pivoxil | 161715-24-8 | MFCD17215369 | 99.7% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB039610MM/1ML
Encompass_Preferred
Tebipenem pivoxil is an orally active antibiotic effective against various pathogenic bacteria. It functions by binding to penicillin-binding protein (PBP), thereby inhibiting cell wall synthesis.
  • Orally active antibiotic
  • Effective against various pathogenic bacteria
  • Inhibits cell wall synthesis
  • High purity of 99.7%
Catalog No. 50-004-07294