Learn More
Apexbio Technology LLC Tebuconazole 107534-96-3 200mg
SDP
Supplier:  Apexbio Technology LLC C6515200
Encompass_Preferred
Tebuconazole (CAS 107534-96-3) is a small-molecule inhibitor targeting CYP51 a key enzyme in fungal sterol biosynthesis It modulates CYP51 activity to regulate sterol production within fungal cells Tebuconazole exerts its biological activity primarily through inhibition of CYP51 In cell-based assays tebuconazole demonstrates inhibitory activity with an IC50 value of 0 9 M against CaCYP51 from Candida albicans and 1 3 M against truncated human 60HsCYP51 Additionally in HepG2 cells tebuconazole induces lipid accumulation and oxidative stress and in mammary epithelial MAC-T cells it reduces proliferation and induces endoplasmic reticulum stress-mediated apoptosis Based on these pharmacological properties tebuconazole holds research potential in studies of fungal sterol biosynthesis cellular stress response pathways and apoptosis mechanisms
Catalog No. 50-000-90686