Learn More
Medchemexpress LLC Tebuconazole-d9 | 1246818-83-6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0852S1MG
Encompass_Preferred
Tebuconazole-d9 is a deuterium-labeled form of Tebuconazole, an agricultural azole fungicide. It functions by inhibiting CYP51 with IC50s of 0.9 μM for Candida albicans CYP51 (CaCYP51) and 1.3 μM for truncated Homo sapiens CYP51 (Δ60HsCYP51).
  • Deuterium-labeled compound
  • Functions as an agricultural azole fungicide
  • Inhibits CYP51
  • Used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development
Catalog No. 50-004-07659