Learn More
Sigma Aldrich Fine Chemicals Biosciences TECHNE DRI-BLOCK HEATER DB-2AG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z3813221EA
Encompass
Techne® Dri-Block® heaterModel DB-2A, AC/DC input 120 V ACAC/DC input: 115 V (AC)usage: two insert blocksfeature: CSA approved: no calibrated analogue dial (±2°C).manufacturer/tradename: Techneavailability: not available in EUtemperature measuring range : 25-100 °Ccapacity: holds 2 blocks
Catalog No. NC2739567