Learn More
Cole-Parmer Techne MCS-104L Biological Stirrer, 4-Position, Large; 110/230 V
Supplier:  Cole-Parmer 2183702
Encompass
The soft-start/stop feature provides slow acceleration and deceleration of the stirrer, avoiding excessive turbulence in the culture media and eliminating cell damage. Use the interval stirring mode during the attachment phase to further reduce agitation of the media or when culturing particularly fragile cells. The stirrers create virtually no heat making them suitable for use in both incubators and cold rooms. The culture vessels incorporate a unique base design which, together with the bulb-ended stirrer, ensures that the cells are lifted into suspension at the lowest possible speeds. This gentle stirring action promotes high cell yields by preventing cell shearing. Order Techne cell cultrure vessels separately.
Catalog No. NC1550681