Learn More
Sigma Aldrich Fine Chemicals Biosciences TechneR digital Dri BlockR1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7413261EA
Encompass
Techne Dri-Block heaters have been providing a safe dry constant temperature source in the laboratory for 50 years. Now Techne are proud to introduce the new 2014 range offering market-leading choice flexibility and specification. Dri-Block heaters are a source of precision temperature control for general heating and incubating applications as well as sensitive analytical procedures particularly suited to microbiology and clinical laboratories. They are designed for precise heating of test-tubes microcentrifuge tubes cuvettes and microtitre plates. Techne Dri-Block heaters cover the temperature range from ambient 5C to 100C or 200C (depending on model) using machined aluminium alloy Dri-Block inserts as the heat transfer medium. Independent indicator lights indicate mains power and heater action.
Catalog No. 50-290-8510