Learn More
Sigma Aldrich Fine Chemicals Biosciences TechneR immersion circulat1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z3811011EA
Encompass
These clip-on units will convert an ordinary water bath into a precision temperature-controlled bath. Each incorporates a thermostatted heater unit circulating pump connectors for optional external circulation and an adjustable over-temperature cutoff. Refrigeration required for operation below ambient (cooling coil available as an option if not supplied standard).All submerged parts are made of stainless steel or high-temperature self-extinguishing plastic.Supplied as circulator only (order bath separately).Thermoregulator bridge mounting plate recommended for use above 150 C.
Catalog No. 50-290-8664