Learn More
Medchemexpress LLC Tectochrysin | 520-28-5 | MFCD00016945 | 99.8% | 268.26 g/mol | C16H12O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY145925MG
Encompass_Preferred
Tectochrysin is a naturally occurring flavonoid research reagent reported to inhibit NF-κB activity. Isolated from Alpinia oxyphylla, it is provided as a high-purity analytical standard suitable for biological and chemical research applications.
  • High purity: 99.8%
  • Identified by CAS 520-28-5 and formula C16H12O4
  • Molecular weight 268.26 g/mol
  • Suitable for in vitro biochemical and cell-based assays
  • Supplied as a 5 mg vial for analytical and preparative use
Catalog No. 50-002-21021