Learn More
Medchemexpress LLC HY-101755 5mg Medchemexpress, Tedatioxetine (hydrobromide) CAS:960151-65-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1017555MG
Encompass_Preferred
Medchemexpress, HY-101755 5mg Tedatioxetine (hydrobromide) CAS:960151-65-9 Tedatioxetine (Lu AA24530) hydrobromide acts as a serotonin and norepinephrine (NE)-preferring triple reuptake inhibitor (TRI) and 5-HT2A, 5-HT2C, 5-HT3 and α1A-adrenergic receptor antagonist
., Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0643