Learn More
TARGETMOL CHEMICALS INC TEDIZOLID 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T174710MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Tedizolid (DA-7157)is a novel oxazolidinone-class antibiotic that inhibits bacterial protein synthesis by binding to the 23S ribosomal RNA of the 50S subunit of the ribosome. purity: 99%
Catalog No. 50-258-1102