Learn More
Apexbio Technology LLC Tedizolid 856866-72-3 50mg
SDP
Supplier:  Apexbio Technology LLC A386350
Encompass_Preferred
Tedizolid (CAS 856866-72-3) is a small-molecule inhibitor targeting bacterial 50S ribosomal subunit and human monoamine oxidase (MAO) It inhibits bacterial protein synthesis by binding to the 50S subunit preventing formation of the functional 70S initiation complex Tedizolid exerts its biological activity primarily through inhibition of protein synthesis and MAO In in vitro studies tedizolid demonstrates antibacterial activity against gram-positive pathogens with an MIC50 of approximately 0 5 g/ml against MRSA and inhibits MAO-A and MAO-B with IC50 values of 8 7 M and 5 7 M respectively Based on these pharmacological properties tedizolid holds research potential in antimicrobial resistance studies and investigations of oxazolidinone pharmacodynamics
Catalog No. 50-000-96835