Learn More
SKC Inc TEDLAR SAMPLE BAG 3L 10/PK
SDP
Supplier:  SKC Inc 23203
Encompass
SKC Tedlar 3 Liter Sample Bag with single polypropylene septum fitting. Fitting combines the hose/valve and the septum holder into one compact fitting, (pkg/10). DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC0327636