Learn More
Waters Corp Waters Tee, MMV, Nano, UPLC System
Supplier:  Waters Corp 289004442
Encompass_Preferred
The nano Tee is utilized within the 2D with Dilution Configuration of the nanoACQUITY UPLC system, functioning as a critical fitting.
  • Utilized in the 2D with dilution configuration.
  • Functions as a fitting.
  • Compatible with UPLC system type.
  • Designed for specific UPLC system.
  • Component for ASM and BSM modules.
  • Replacement is as required.
Catalog No. NC1332729