Learn More
Electron Microscopy Sciences Teeny Writer, Black, Extra Fine (0.4 mm)
SDP
Supplier:  Electron Microscopy Sciences 62052B
Encompass_Preferred
Teeny Writers are perfect general use lab markers. The original Teeny Writer has an extra fine tip. Both are ideal for writing on Tough-Tags and Tough-Spots. Use for labeling samples stored at room temperature and not exposed to chemicals and/or caustic agents.
Color: Black
Size: 0.4 mm
Catalog No. 50-346-24