Learn More
World Precision Instrument TEER Electrode Chloride Solution
SDP
Supplier:  World Precision Instrument TEERCL
Encompass
  • Solution to chloride your TEER measuring system electrodes.
  • It is recommended chloriding your electrode every 2-3 days if used frequently.
  • 1 bottle, Volume of 20 mL
Catalog No. 50-253-8487