Learn More
Strem, An Ascensus Company CAS# 1115-99-7. 100g. Triethylgallium, elec. gr. (99.9999%-Ga) PURATREM. MFCD00015094
SDP
Supplier:  Strem, An Ascensus Company 981862100G
Encompass
CAS# 1115-99-7. 100g. Triethylgallium, elec. gr. (99.9999%-Ga) PURATREM. MFCD00015094. Molecular Weight: 156.91. Molecular Formula: (C2H5)3Ga. Color/form: colorless liq. Strem# 98-1862. http://www.strem.com/catalog/v/98-1862/
Catalog No. 50-204-5447
Only null left
Add to Cart
Request a Quote